Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prostein Mouse anti-Human, Clone: 5E12E2, Proteintech
SDP

Catalog No. 501736430 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6430 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6430 Supplier Proteintech Group Inc Supplier No. 603431IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Prostein is a novel prostate-specific protein expressed in normal and maligt prostate tissues. This protein appears to be a type IIIa plasma membrane protein with a cleavable signal peptide and 11 transmembrane-spanning regions. Prostein expression is androgen responsive since treatment of LNCaP cells with androgen up-regulates prostein message and protein expression levels. Thus, prostein is a prostate-specific marker whose detection and quantitation may be useful in diagnosing and monitoring prostate cancer.

Specifications

Antigen Prostein
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 5E12E2
Concentration 1.47 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SLC45A3
Gene Accession No. Q96JT2
Gene Alias IPCA 2, IPCA 6, IPCA 8, IPCA6, PCANAP2, PCANAP6, PCANAP8, Prostein, PRST, SLC45A3
Gene Symbols SLC45A3
Host Species Mouse
Immunogen Prostein Fusion Protein Ag5463
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85414
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.