Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Prostein Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553914
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553914 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553914 Supplier Invitrogen™ Supplier No. PA553914
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AEEAALGPTE PAEGLSAPSL SPHCCPCRAR LAFRNLGALL PRLHQLCCRM P Highest antigen sequence identity to the following orthologs: Mouse - 69%, Rat - 75%.

Prostein is a novel prostate-specific protein expressed in normal and malignant prostate tissues. This protein appears to be a type IIIa plasma membrane protein with a cleavable signal peptide and 11 transmembrane-spanning regions. Prostein expression is androgen responsive since treatment of LNCaP cells with androgen up-regulates prostein message and protein expression levels. Thus, prostein is a prostate-specific marker whose detection and quantitation may be useful in diagnosing and monitoring prostate cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Prostein
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC45A3
Gene Accession No. Q96JT2
Gene Alias 2210413P12Rik; AU023994; AU043764; IPCA-2; IPCA6; IPCA-6; IPCA-8; PCANAP2; PCANAP6; PCANAP8; prostate cancer associated protein 2; prostate cancer associated protein 6; prostate cancer associated protein 8; prostate cancer-associated gene 2; prostate cancer-associated gene 6; prostate cancer-associated gene 8; prostate cancer-associated protein 6; Prostein; PRST; RGD1309764; SLC45A3; Solute carrier family 45 member 3; solute carrier family 45, member 3
Gene Symbols SLC45A3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Prostein. Recombinant protein control fragment (Product #RP-92527).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85414
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.