Learn More
Prostein Recombinant Rabbit Monoclonal Antibody (ZR9), RAbMono™, Zeta
Rabbit Recombinant Monoclonal Antibody Cocktail
$399.84
Specifications
Antigen | Prostein |
---|---|
Clone | ZR9 |
Concentration | 200 μg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-221-5773
![]() |
ZETA CORPORATION
Z2006RS |
500 μL |
Each of 1 for $399.84
|
|
|||||
Description
A recommended positive control tissue for this product is Prostate, however positive controls are not limited to this tissue type. The primary antibody is intended for laboratory professional use in the detection of the corresponding protein in formalin-fixed, paraffin-embedded tissue stained in manual qualitative immunohistochemistry (IHC) testing. This antibody is intended to be used after the primary diagnosis of tumor has been made by conventional histopathology using non-immunological histochemical stains.
Prostein Recombinant Monoclonal antibody specifically detects Prostein in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Prostein | |
200 μg/mL | |
Monoclonal | |
Liquid | |
RUO | |
tris with BSA, NP-40 and <0.1% sodium azide | |
2210413P12Rik; AU023994; AU043764; IPCA-2; IPCA6; IPCA-6; IPCA-8; PCANAP2; PCANAP6; PCANAP8; prostate cancer associated protein 2; prostate cancer associated protein 6; prostate cancer associated protein 8; prostate cancer-associated gene 2; prostate cancer-associated gene 6; prostate cancer-associated gene 8; prostate cancer-associated protein 6; Prostein; PRST; RGD1309764; SLC45A3; Solute carrier family 45 member 3; solute carrier family 45, member 3 | |
Synthesized peptides to the N-terminus of human protein | |
Primary | |
2-8°C | |
SLC45A3 |
ZR9 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
Q96JT2 | |
85414 | |
IgG | |
Protein A | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.