Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Prosurfactant Protein C Antibody (5E6A9) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$560.50
Specifications
Antigen | Prosurfactant Protein C |
---|---|
Clone | 5E6A9 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000 |
Applications | Western Blot, ELISA |
Description
Prosurfactant Protein C Monoclonal specifically detects Prosurfactant Protein C in Human samples. It is validated for Western Blot, ELISA.Specifications
Prosurfactant Protein C | |
1 mg/mL | |
Western Blot, ELISA | |
Unconjugated | |
RUO | |
PBS with 0.05% Sodium Azide | |
6440 | |
Purified recombinant fragment of human SFTPC (AA: 60-180 ) expressed in E. Coli. | |
Primary | |
21 kDa |
5E6A9 | |
Western Blot 1:500 - 1:2000, ELISA 1:10000 | |
Monoclonal | |
Mouse | |
Human | |
PSP-C, pulmonary surfactant-associated protein C, Pulmonary surfactant-associated proteolipid SPL(Val), SFTP2, SFTP2pulmonary surfactant apoprotein-2 SP-C, SMDP2surfactant, pulmonary-associated protein C, SP-CSP5, surfactant protein C | |
SFTPC | |
IgG1 | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title