Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ proTAME
GREENER_CHOICESDP

Catalog No. 77341 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
77-341 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 77-341 Supplier Tocris Bioscience™ Supplier No. 7734/1
Add to Cart
Add to Cart
This item is not returnable. View return policy

Cell-permeable APC/C inhibitor

ProTAME is a cell-permeable APC/C (anaphase-promoting complex/cyclosome) inhibitor. It is converted to TAME (Tosyl-L-Arginine Methyl Ester) by intracellular esterases. It induces cell cycle arrest in metaphase followed by cell death.

Specifications

Chemical Name or Material Methyl (2S)-2-[[(4-methylphenyl)sulfonyl]amino]-9,13-dioxo-14-phenyl-7-[[[[(2-phenylacetyl)oxy]methoxy]carbonyl]amino]-10,12-dioxa-6,8-diazatetradec-6-enoate
CAS 1362911-19-0
Quantity 1 mg
Target APC
Molecular Formula C34H38N4O12S
Purity 90%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.