Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Protamine 2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP303223100UL
Description
Protamine 2 Polyclonal specifically detects Protamine 2 in Human samples. It is validated for Western Blot.Specifications
Protamine 2 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
5620 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
Unconjugated | |
cancer/testis antigen family 94, member 2, CT94.2, FLJ27447, protamine 2, protamine-2, Sperm histone P2, Sperm protamine P2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-102 of human Protamine 2 (NP_002753.2). | |
100 μL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction