Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Protamine sulfate
SDP

Catalog No. 882210 Shop All Novus Biologicals Products
Change view
Click to view available options
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
88-221-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 88-221-0 Supplier Tocris Bioscience™ Supplier No. 8822/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Transduction efficiency enhancer, also apelin receptor antagonist

Protamine sulfate is a cationic peptide that enhances viral vector transduction efficiency in gene therapy applications. By neutralizing charge repulsion between viral particles and the cell surface, Protamine sulfate promotes vector attachment and uptake, leading to increased transduction efficiency. It is commonly used in combination with Polybrene (Cat. No. 7711) to enhance the delivery of adeno-associated virus (AAV) and lentiviral vectors. Protamine sulfate is widely applied in gene delivery workflows, including stem cell engineering and gene-modified cell therapy development. It is also an antagonist of apelin receptor. For more information on how to prepare Protamine sulfate for transfection enhancement, please see our protocol . For more information on how to prepare Protamine sulfate for enhanced-viral-transduction of mammalian cells, please see our protocol . Protamine sulfate is supplied as a lyophilized solid in a convenient, ready-to-reconstitute vial size. A stock solution can be prepared by adding 1 mL of sterile ultra-pure water directly to the vial (final stock concentration: 10 mg/mL).

Specifications

CAS 53597-25-4
Quantity 10 mg
Target Viral Transduction Enhancers
Purity 0.9
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.