Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Protease-Activated Receptor-4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102297
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102297 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102297 Supplier Invitrogen™ Supplier No. PA5102297
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PAR4.

Coagulation factor II (thrombin) receptor-like 3 (F2RL3) is a member of the large family of 7-transmembrane-region receptors that couple to guanosine-nucleotide-binding proteins. F2RL3 is also a member of the protease-activated receptor family. F2RL3 is activated by proteolytic cleavage of its extracellular amino terminus. The new amino terminus functions as a tethered ligand and activates the receptor. F2RL3 is activated by thrombin and trypsin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Protease-Activated Receptor-4
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene F2RL3
Gene Accession No. O88634, Q920E0, Q96RI0
Gene Alias coagulation factor II (thrombin) receptor-like 3; coagulation factor II receptor-like 3; F2R like thrombin/trypsin receptor 3; F2rl3; PAR4; PAR-4; protease-activated receptor 4; protease-activated receptor-4; proteinase-activated receptor 4; Thrombin receptor-like 3
Gene Symbols F2RL3
Host Species Rabbit
Immunogen A synthesized peptide derived from human F2RL3(Accession Q96RI0), corresponding to amino acid residues G35-G85.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116498, 14065, 9002
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.