Learn More
Tocris Bioscience™ Protease Inhibitor Cocktail I
GREENER_CHOICESDP
Catalog No. 55001ML Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mL
Catalog No. Quantity
55001ML 1 mL
1 options
This item is not returnable. View return policy
Catalog No. 55001ML Supplier Tocris Bioscience™ Supplier No. 5500/1ML
Add to Cart
Add to Cart
This item is not returnable. View return policy
Protease inhibitor cocktail for mammalian cells
Protease Inhibitor Cocktail I is a blend of protease inhibitors optimized for mammalian cells. Contains the following inhibitors: AEBSF HCl (100 mM), Aprotinin (80 μM), Bestatin (5 mM), E-64 (1.5 mM), Leupeptin (2 mM) and Pepstatin (1 mM). Cocktail is provided as 1 ml of 100X concentrate in DMSO.
Specifications
Quantity 1 mL
Target Buffers, Chelators and Reagents