Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Protease Inhibitor Cocktails
GSA_VA

Catalog No. PI78415
Click to view available options
Quantity:
1 mL
2 mL
Product Type:
Protease Inhibitor Cocktail
Protease Inhibitor Cocktail, EDTA-Free

Protect your sample from proteolytic degradation during cell lysis with these broad-spectrum protease inhibitor cocktails.

Provide broad-spectrum protection against proteolytic degradation in cell lysis procedures with Thermo Scientific™ Protease Inhibitor Cocktails.

Both ready-to-use formulations inhibit most serine, cysteine, and calpain proteases that cause general degradation of proteins in cell extracts. Metalloproteases are inhibited as well. The EDTA-free formulation is ideal for preparing samples that will be analyzed by 2D gel electrophoresis. The refrigerator-stable, 100X liquid solutions are easy to use: simply add the appropriate volume of cocktail to the sample (10μL per 1mL of cell extract) and continue with your procedure. The Protease Inhibitor can be used with animal cells and tissues, bacteria, plants and yeast.

Highlights:

  • Ready-to-use, 100X liquid cocktail
  • Available with EDTA (supplied in separate vial) or as an EDTA-free formulation
  • Effective for suppressing proteolytic activity in detergent-based cell lysis reagents
  • No loss of valuable freezer space; cocktail is stable at 4°C

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Formulation Proprietary components in DMSO
Product Type Protease Inhibitor Cocktail, EDTA-Free
Form Liquid
Sufficient For 100 mL of Sample
Quantity 1 mL
Composition Proprietary formulation
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.