Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega ProteaseMAX™ Surfactant, Trypsin Enhancer

Catalog No. PRV2071 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
5 x 1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV2071 1 mg
PRV2072 5 x 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV2071 Supplier Promega Supplier No. V2071
Add to Cart
Add to Cart

ProteaseMAX Surfactant, Trypsin Enhancer, enhances the enzymatic performance of Trypsin, Chymotrypsin and Lys-C in digestion reactions.

  • Improved In-Gel and In-Solution Protein Digestion
  • Enhances enzymatic performance of trypsin, chymotrypsin and Lys-C
  • Saves time and labor when used with in-gel protein digestion
  • Complete digestion in 1 hour

Specifications

Formulation Lyophilized
Includes 1mg ProteaseMAX™ Surfactant, lyophilized
Product Type Trypsin Enhancer
Content And Storage -30°C to -10°C
For Use With (Application) Protein isolation and purification
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.