Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Proteasome 19S 10B Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299616MFV500
Description
Proteasome 19S 10B Polyclonal antibody specifically detects Proteasome 19S 10B in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
Proteasome 19S 10B | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
26S proteasome regulatory subunit RPN1, 26S proteasome regulatory subunit S2, MGC14274,55.11 protein, P97, proteasome (prosome, macropain) 26S subunit, non-ATPase, 2, Protein 55.11, Rpn1, S226S proteasome non-ATPase regulatory subunit 2, TNFR-associated protein 2, TRAP226S proteasome subunit p97, tumor necrosis factor receptor-associated protein 2, Tumor necrosis factor type 1 receptor-associated protein 2 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human Proteasome 19S 10B. | |
0.1 mL | |
Cell Cycle and Replication, Neuroscience, Ubiquitin Proteasome Pathway | |
5708 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunoprecipitation | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction