Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Proteasome 19S 10B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB300532
Description
Proteasome 19S 10B Polyclonal specifically detects Proteasome 19S 10B in Human, Mouse, Rat, Canine, Hamster, Primate samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Proteasome 19S 10B | |
Polyclonal | |
Western Blot 1:1000 | |
26S proteasome regulatory subunit RPN1, 26S proteasome regulatory subunit S2, MGC14274,55.11 protein, P97, proteasome (prosome, macropain) 26S subunit, non-ATPase, 2, Protein 55.11, Rpn1, S226S proteasome non-ATPase regulatory subunit 2, TNFR-associated protein 2, TRAP226S proteasome subunit p97, tumor necrosis factor receptor-associated protein 2, Tumor necrosis factor type 1 receptor-associated protein 2 | |
Rabbit | |
Ammonium sulfate precipitation | |
RUO | |
Primary | |
Detects proteasome 19S subunit 10B. | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
Unconjugated | |
PBS with 1 mg/ml BSA with 0.05% Sodium Azide | |
PSMD2 | |
Synthetic Peptide: C A(278) T N R P D T L D P A L L R P G R(294) | |
100 μL | |
Cell Cycle and Replication, Neuroscience, Ubiquitin Proteasome Pathway | |
5708 | |
Human, Mouse, Rat, Canine | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction