Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Proteasome 19S S3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | Proteasome 19S S3 |
---|---|
Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
Proteasome 19S S3 Polyclonal specifically detects Proteasome 19S S3 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Proteasome 19S S3 | |
Polyclonal | |
Rabbit | |
Neuroscience, Ubiquitin Proteasome Pathway | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
26S proteasome regulatory subunit RPN3, 26S proteasome regulatory subunit S3, P58, proteasome (prosome, macropain) 26S subunit, non-ATPase, 3, Proteasome subunit p58, RPN3, S3, 26S proteasome non-ATPase regulatory subunit 3, tissue specific transplantation antigen 2, TSTA2 | |
PSMD3 | |
IgG | |
Affinity Purified |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
O43242 | |
5709 | |
This antibody was developed against a recombinant protein corresponding to amino acids: SINHEKGYVQSKEMIDIYSTREPQLAFHQRISFCLDIHNMSVKAMRFPPKSYNKDLESAEERREREQQDLEFAKEMAEDDD | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title