Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Proteasome 19S S3 Antibody (OTI2A2), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273636V
Description
Proteasome 19S S3 Monoclonal specifically detects Proteasome 19S S3 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry.Specifications
Proteasome 19S S3 | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PSMD3 | |
Full length recombinant protein of human Proteasome 19S S3 (NP_002800) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Canine, Primate, Monkey | |
IgG2a |
Western Blot, Flow Cytometry | |
OTI2A2 | |
Western Blot, Flow Cytometry | |
26S proteasome regulatory subunit RPN3, 26S proteasome regulatory subunit S3, P58, proteasome (prosome, macropain) 26S subunit, non-ATPase, 3, Proteasome subunit p58, RPN3, S3, 26S proteasome non-ATPase regulatory subunit 3, tissue specific transplantation antigen 2, TSTA2 | |
Mouse | |
Affinity Purified | |
Neuroscience, Ubiquitin Proteasome Pathway | |
5709.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction