Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Proteasome 19S S7 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | Proteasome 19S S7 |
---|---|
Dilution | Western Blot 0.4 ug/ml, Simple Western, Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Proteasome 19S S7 Polyclonal specifically detects Proteasome 19S S7 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Proteasome 19S S7 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
26S protease regulatory subunit 7, 26S proteasome AAA-ATPase subunit RPT1, MGC3004, MSS1ATPase, 2, proteasome (prosome, macropain) 26S subunit, ATPase, 2, Protein MSS1, putative protein product of Nbla10058 | |
PSMC2 | |
IgG | |
Affinity Purified | |
Specificity of human Proteasome 19S S7 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot 0.4 ug/ml, Simple Western, Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
Polyclonal | |
Rabbit | |
Neuroscience, Ubiquitin Proteasome Pathway | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
5701 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:RKTKEDEKDDKPIRALDEGDIALLKTYGQSTYSRQIKQVEDDIQQLLKKINELTGIKESDTGLAPPALWDLAA | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title