Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Proteasome 19S S7 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297271PCP
Description
Proteasome 19S S7 Polyclonal antibody specifically detects Proteasome 19S S7 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
Proteasome 19S S7 | |
Polyclonal | |
Immunocytochemistry/ Immunofluorescence | |
26S protease regulatory subunit 7, 26S proteasome AAA-ATPase subunit RPT1, MGC3004, MSS1ATPase, 2, proteasome (prosome, macropain) 26S subunit, ATPase, 2, Protein MSS1, putative protein product of Nbla10058 | |
Produced in rabbits immunized with E. coli-derived Human Proteasome 19S S7 fragment. | |
0.1 mL | |
Neuroscience, Ubiquitin Proteasome Pathway | |
5701 | |
Store at 4C in the dark. | |
IgG |
Immunocytochemistry | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction