Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Proteasome 20S alpha 3 Antibody (SR1471), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $423.00
Specifications
Antigen | Proteasome 20S alpha 3 |
---|---|
Clone | SR1471 |
Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation |
Classification | Monoclonal |
Description
Proteasome 20S alpha 3 Monoclonal antibody specifically detects Proteasome 20S alpha 3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, ImmunoprecipitationSpecifications
Proteasome 20S alpha 3 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
EC 3.4.25.1, HC8MGC32631, Macropain subunit C8, MGC12306, Multicatalytic endopeptidase complex subunit C8, proteasome (prosome, macropain) subunit, alpha type, 3, Proteasome component C8, proteasome subunit alpha type-3, proteasome subunit C8, PSC3, PSC8 | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1471 | |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Stem Cell Markers | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
5684 | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title