Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Proteasome 20S alpha 3 Rabbit anti-Human, Mouse, Rat, Clone: 7X8W9, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP31655120UL
This item is not returnable.
View return policy
Description
Proteasome 20S alpha 3 Monoclonal antibody specifically detects Proteasome 20S alpha 3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Proteasome 20S alpha 3 | |
Monoclonal | |
Unconjugated | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
7X8W9 | |
Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
EC 3.4.25.1, HC8MGC32631, Macropain subunit C8, MGC12306, Multicatalytic endopeptidase complex subunit C8, proteasome (prosome, macropain) subunit, alpha type, 3, Proteasome component C8, proteasome subunit alpha type-3, proteasome subunit C8, PSC3, PSC8 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 21-150 of human Proteasome 20S alpha 3 (P25788). | |
20 μg | |
Stem Cell Markers | |
5684 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction