Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Proteasome 20S alpha 5 Antibody (AH1.1), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP276657JF549
Description
Proteasome 20S alpha 5 Monoclonal specifically detects Proteasome 20S alpha 5 in Human, Xenopus samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Proteasome 20S alpha 5 | |
Monoclonal | |
Janelia Fluor 549 | |
50 mM sodium borate with 0.05% sodium azide | |
PSMA5 | |
Sub-unit 5a | |
0.1 mL | |
Primary | |
Human, Xenopus species | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
AH1.1 | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
EC 3.4.25.1, FLJ42315, macropain subunit zeta, Macropain zeta chain, MGC117302, MGC125802, MGC125803, MGC125804, Multicatalytic endopeptidase complex zeta chain, proteasome (prosome, macropain) subunit, alpha type, 5, proteasome alpha 5 subunit, proteasome component 5, proteasome subunit alpha type-5, proteasome subunit zeta, Proteasome zeta chain, PSC5, ZETA | |
Mouse | |
Protein A purified | |
Core ESC Like Genes, Stem Cell Markers | |
5686 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction