Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Proteasome 20S alpha 6 Antibody (OTI3B8), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273642FR
Description
Proteasome 20S alpha 6 Monoclonal specifically detects Proteasome 20S alpha 6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Proteasome 20S alpha 6 | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
PSMA6 | |
Full length recombinant protein of human Proteasome 20S alpha 6 (NP_002782) produced in E.coli. | |
0.1 mL | |
5687.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI3B8 | |
Western Blot, Immunohistochemistry | |
EC 3.4.25.1, IOTA, Macropain iota chain, macropain subunit iota, MGC22756, MGC2333, MGC23846, Multicatalytic endopeptidase complex iota chain, P27K, p27KPROS-27, PROS2727 kDa prosomal protein, prosomal P27K protein, proteasome (prosome, macropain) subunit, alpha type, 6, Proteasome iota chain, proteasome subunit alpha type-6, proteasome subunit iota | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction