Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Proteasome 20S beta 6 Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP314763H
Description
Proteasome 20S beta 6 Polyclonal antibody specifically detects Proteasome 20S beta 6 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Proteasome 20S beta 6 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
DELTA, EC 3.4.25.1, LMPY, Macropain delta chain, Multicatalytic endopeptidase complex delta chain, proteasome (prosome, macropain) subunit, beta type, 6, proteasome catalytic subunit 1, Proteasome delta chain, proteasome subunit beta 6, proteasome subunit beta type-6, proteasome subunit delta, Proteasome subunit Y, PSY large multifunctional protease Y, YMGC5169 | |
E. coli-derived Human PSMB6 fragment | |
0.2 mL | |
Core ESC Like Genes, Neuroscience, Stem Cell Markers, Ubiquitin Proteasome Pathway | |
5694 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
HRP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction