Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Proteasome 20S C2 Rabbit anti-Human, Mouse, Rat, Clone: JB89-30, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP275628
Description
PSMA1 Monoclonal specifically detects PSMA1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Proteasome 20S C2 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation, Immunohistochemistry-Paraffin 1:50-1:200 | |
EC 3.4.25.1, HC2Proteasome nu chain, Macropain subunit C2, macropain subunit nu, MGC14542, MGC14575, MGC14751, MGC1667, MGC21459, MGC22853, MGC23915, Multicatalytic endopeptidase complex subunit C2, NU30 kDa prosomal protein, PROS-30, PROS30Proteasome component C2, proteasome (prosome, macropain) subunit, alpha type, 1, proteasome subunit alpha type-1, proteasome subunit nu, proteasome subunit, alpha-type, 1, protein P30-33K, PSC2 | |
Rabbit | |
30 kDa | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
JB89-30 | |
Unconjugated | |
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide | |
PSMA1 | |
Synthetic peptide within Human PSMA1 aa 214-263/ 263. (SwissProt: P25786 Human; SwissProt: Q9R1P4 Mouse; SwissProt: P18420 Rat) | |
Protein A purified | |
Neuroscience, Ubiquitin Proteasome Pathway | |
5682.0 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction