Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Proteasome 26S S5 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1203319
Description
Proteasome 26S S5 Polyclonal specifically detects Proteasome 26S S5 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| Proteasome 26S S5 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| PSMD5 | |
| Synthetic Peptide: L(485) S E G P Y Y V K P V S T T A V E G A E(504) | |
| 100 μg | |
| Neuroscience, Ubiquitin Proteasome Pathway | |
| 5711 | |
| Human | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1.0 ug/ml | |
| KIAA007226S proteasome non-ATPase regulatory subunit 5, proteasome (prosome, macropain) 26S subunit, non-ATPase, 5,26S proteasome subunit S5B, S5BMGC23145,26S protease subunit S5 basic | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| NB120-3319 immunizing peptide corresponds to amino acid residues 485-504 from human proteasome 19S subunit S5b protein. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction