Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Proteasome subunit beta type 4 Antibody (OTI2C9), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273655G
Description
Proteasome subunit beta type 4 Monoclonal specifically detects Proteasome subunit beta type 4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Proteasome subunit beta type 4 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PSMB4 | |
Full length recombinant protein of human Proteasome subunit beta type 4 (NP_002787) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI2C9 | |
Western Blot, Immunohistochemistry | |
EC 3.4.25.1, HN3, hsBPROS26, HsN3, Macropain beta chain, Multicatalytic endopeptidase complex beta chain, PROS26PROS-26, proteasome (prosome, macropain) subunit, beta type, 4, Proteasome beta chain, Proteasome chain 3, proteasome subunit beta type-4, proteasome subunit HsN3, proteasome subunit, beta type, 4,26 kDa prosomal protein | |
Mouse | |
Affinity Purified | |
Stem Cell Markers | |
5692.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction