Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Protector™Acid Storage Cabinets
GSA_VASDP

Catalog No. 16080305 Shop All Labconco Corporation Products
Change view
Click to view available options
Door Type:
Single Left Hinged Door
Single Right Hinged Door
Width (English):
18 in.
24 in.
Height (English) Base:
31.5 to 32.75 in.
35.5 to 36.75 in.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Door Type Width (English) Height (English) Base
16-080-305 Single Left Hinged Door 18 in. 35.5 to 36.75 in.
16-080-306 Single Right Hinged Door 24 in. 31.5 to 32.75 in.
16-080-309 Single Left Hinged Door 18 in. 31.5 to 32.75 in.
16-080-308 Single Left Hinged Door 24 in. 31.5 to 32.75 in.
16-080-307 Single Right Hinged Door 18 in. 31.5 to 32.75 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 16-080-305 Supplier Labconco™ Supplier No. 9901600
Add to Cart
Add to Cart

Includes

Vacuum-formed PVC liner tray, two rear vent connection cutouts for venting the cabinet into a fume hood, four leveling feet and one 8 in. filler panel to increase cabinet depth from 22 to 30 in.

Support Labconco fume hoods and safely store and vent acids and other corrosive liquids

  • Corrosion-resistant polyethylene-lined interior
  • Manual-closing, non-locking door(s)

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Product Type Storage Cabinet
Color Light Gray
Weight (Metric) 40.82 kg
Weight (English) 90 lb.
Includes Vacuum formed PVC liner tray, 2 rear vent connection cut outs for venting the cabinet into a fume hood, 4 leveling feet and one 8 in. filler panel to increase cabinet depth from 22 to 30in.
Load Bearing Capacity (Metric) 362.8 kg
Width (English) 18 in.
Load Bearing Capacity (English) 800 lb.
Height (Metric) Base 90.17 to 93.34 cm
Depth (English) 22 in.
Depth (Metric) 55.88 cm
Door Type Single Left Hinged Door
For Use With (Application) Ideal for storage and venting of small laboratory instrument and mild acid and other reagent
Height (English) Base 35.5 to 36.75 in.
Material Steel
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.