Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Protein A Mag Sepharose™ Xtra

Catalog No. 45002510 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
2 x 1 mL
5 x 1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-002-510 2 x 1 mL
45-002-511 5 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-002-510 Supplier Cytiva Supplier No. 28967056
Add to Cart
Add to Cart

Magnetic beads are designed for rapid, small-scale purification and screening of monoclonal and polyclonal antibodies from serum and cell supernatants

Used as a separation tool for handling the beads in microcentrifuge tubes, up to six samples can be processed in parallel and can be easily screen a larger number of samples in parallel with high throughput on a robotic device.

  • Visible and dense Sepharose-based magnetic beads with protein A ligand allow easy spotting and collection of bound target protein
  • Non-adherent beads eliminate smearing effects and aggregate formation
  • Can be used without detergents
  • Simple capture of target protein in small or large sample volumes (low microliter to high milliliter scale)
  • Provides high purity and yield

∽350μg purified human IgG can be obtained from each purification run
Provides robust parallel screening of antibodies with high reproducibility
Matrix: paramagnetic, spherical, highly cross-linked agarose particles
Ligand: native protein A
Binding capacity: 27mg human IgG/mL gel
Operating temp.: room temperature
Stability: stable

  • Small-scale purification
  • Antibody screening
  • Antibody purification

TRUSTED_SUSTAINABILITY

Specifications

Binding Capacity 27mg human IgG/mL gel
Quantity 2 x 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.