Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Protein A Polyclonal Antibody, FITC, Rockland™
SDP

Catalog No. 501990331
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0331 25 μL
50-199-0330 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-0331 Supplier Rockland Immunochemicals Supplier No. 200102077S
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-fluorescein, anti-Goat Serum, and purified and partially purified Protein A.

Protein A is an Orphan-U GPCR with an unknown ligand. This gene was identified from genomic analysis of a gene-dense region at human chromosome 12p13. It appears to be expressed mainly in the brain. However, its function is not known. Three alternatively spliced variants have been described; the full length of the third variant is yet to be determined.

Specifications

Antigen Protein A
Applications Flow Cytometry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate FITC
Formulation 0.02M potassium phosphate with 0.15M NaCl, 10 mg/mL BSA and 0.01% sodium azide; pH 7.2
Host Species Goat
Immunogen Protein A (Staphylococcus aureus)
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.