Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Protein A Polyclonal Antibody, Invitrogen™
Chicken Polyclonal Antibody
Supplier: Thermo Scientific PA172062
Description
PA1-72062 detects Protein A in bacteria samples. PA1-72062 has been successfully used in ELISA and western blot procedures. The PA1-72062 immunogen is from purified recombit protein A.
Protein A is 56 kDA surface protein found in the cell wall of Staphylococcus aureus. Protein A has a high affinity to Human IgG, Mouse IgG and others and is therefore suitable for purification and detection of immunglobulins.
Specifications
Protein A | |
Polyclonal | |
Unconjugated | |
Purified recombit protein A. | |
500 μg | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
2 mg/mL | |
Chicken | |
Affinity chromatography | |
RUO | |
Bacteria | |
Antibody | |
IgY |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction