Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novex™ Protein A - Sepharose™ 4B

Catalog No. p-7076184
Encompass_Preferred
Click to view available options
Quantity:
5 mL
25 mL
1 L

Protein A - Sepharose™ 4B

This product is prepared by covalently coupling highly purified (>98% by SDS-PAGE) Staph. aureus Cowan strain 1 Protein A to Sepharose™ 4B.

This product is supplied as a 50% suspension consisting of Protein A-Sepharose™ 4B and PBS (10 mM phosphate-buffered saline, pH 7.4, containing 0.1% Proclin).

Applications: Immunoglobulin Affinity Purification, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Product Type Protein A Sepharose
Product Line Sepharose™
Column Type Affinity
Stationary Phase Protein A
Content And Storage This product is supplied as a 50% suspension (5 ml Protein A-Sepharose™ 4B + 5 ml PBS) Wet Ice
Quantity 5 mL
Form Suspension
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.