Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Protein A Sepharose™ CL-4B Medium
GSA_VA

Catalog No. 45000171 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1.5 g
25 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-000-171 500 mL
45-000-143 1.5 g
45-000-172 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 45-000-171 Supplier Cytiva Supplier No. 17096302
Add to Cart
Add to Cart
Request a Quote

For immunoglobin purification and fractionation. Cytiva Protein A Sepharose™ CL-4B Medium is also well suited for immunoprecipitation.

  • Strong binding to Fc region of IgG
  • Suitable for polyclonal antibody purification
  • High capacity—20mg human IgG/mL
  • pH stability Cleaning-in-Place (CIP): 2 to 10
  • Stable in commonly used aqueous buffers, 6M guanidine-HCl, 8M urea

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Binding Capacity 20mg Human IgG/mL
Ligand Concentration 3mg Protein A/mL Medium
Matrix 4% Cross Linked Agarose
Particle Size 90 μm
pH 2 to 10
Quantity 500 mL
Content And Storage 2°C to 8°C
For Use With (Application) For immunoglobin purification and fractionation and is suitable for immunoprecipitation procedures
Ligand Native Protein A
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.