Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SouthernBiotech™ Protein A-Unlabeled
Shop All Southern Biotechnology Associates Products

Click to view available options
Quantity:
1 mg
5 mg
Specifications
Specifications
Conjugate | Unconjugated |
Formulation | Supplied lyophilized from distilled water; No BSA, preservatives or amine-containing buffer salts added |
For Use With (Application) | ELISA, IP, WB, Purification |
Content And Storage | -20°C |
Quantity | 5 mg |
For Research Use only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction