Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SouthernBiotech™ Protein A-Unlabeled
SDP

Catalog No. p-7139061 Shop All Southern Biotechnology Associates Products
Click to view available options
Quantity:
1 mg
5 mg

SouthernBiotech binding proteins are useful for immunoprecipitation, purification, and ELISA applications.

Specifications

Conjugate Unconjugated
Formulation Supplied lyophilized from distilled water; No BSA, preservatives or amine-containing buffer salts added
For Use With (Application) ELISA, IP, WB, Purification
Content And Storage -20°C
Quantity 5 mg

For Research Use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.