Learn More
Invitrogen™ Protein APC Recombinant Rabbit Monoclonal Antibody (8X7Q1)

Description
Protein APC Recombinant Monoclonal Antibody for ELISA, IP, Western Blot
Specifications
Specifications
| Antigen | Protein APC |
| Applications | ELISA, Immunoprecipitation, Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 8X7Q1 |
| Concentration | 1.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, 0.05% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | Apc |
| Gene Accession No. | P25054, P70478, Q61315 |
| Gene Alias | adenomatosis polyposis coli; adenomatosis polyposis coli tumor suppressor; adenomatous polyposis coli; adenomatous polyposis coli protein; AI047805; APC; APC, WNT signaling pathway regulator; AU020952; AW124434; BTPS2; CC1; Deleted in polyposis 2.5; DP2; DP2.5; DP3; FAP; FPC; GS; mAPC; Min; multiple intestinal neoplasia; PPP1R46; Protein APC; protein phosphatase 1, regulatory subunit 46; RATAPC; truncated adenomatosis polyposis coli; WNT signaling pathway regulator |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.