Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Protein APC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596213
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596213 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596213 Supplier Invitrogen™ Supplier No. PA596213
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QSKENQVSAK GTWRKIKENE FSPTNSTSQT VSSGATNGAE SKTLIYQMAP AVSKTEDVWV RIEDCPINNP RSGRSPTGNT PPVIDSVSEK ANPNIKDSKD NQAKQNVGNG SVPMRTVGLE NRLNSFIQVD APDQKGTEIK PGQNNPVPVS ETNESSIVER T Positive Samples: HeLa; Cellular Location: Cell junction, Cell membrane, Cell projection, Cytoplasm, adherens junction, cytoskeleton, lamellipodium, ruffle membrane.

This gene encodes a tumor suppressor protein that acts as an antagonist of the Wnt signaling pathway. It is also involved in other processes including cell migration and adhesion, transcriptional activation, and apoptosis. Defects in this gene cause familial adenomatous polyposis (FAP), an autosomal dominant pre-malignant disease that usually progresses to malignancy. Disease-associated mutations tend to be clustered in a small region designated the mutation cluster region (MCR) and result in a truncated protein product.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Protein APC
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 1.99 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Apc
Gene Accession No. P25054, P70478, Q61315
Gene Alias adenomatosis polyposis coli; adenomatosis polyposis coli tumor suppressor; adenomatous polyposis coli; adenomatous polyposis coli protein; AI047805; APC; APC, WNT signaling pathway regulator; AU020952; AW124434; BTPS2; CC1; Deleted in polyposis 2.5; DP2; DP2.5; DP3; FAP; FPC; GS; mAPC; Min; multiple intestinal neoplasia; PPP1R46; Protein APC; protein phosphatase 1, regulatory subunit 46; RATAPC; truncated adenomatosis polyposis coli; WNT signaling pathway regulator
Gene Symbols Apc
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 2600-2760 of human APC (NP_0000292).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11789, 24205, 324
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.