Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Protein C (Active) Monoclonal Antibody (PC107)
GREENER_CHOICE

Catalog No. FERMA549088
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA549088 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA549088 Supplier Invitrogen™ Supplier No. MA549088
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The monoclonal antibody PC107 is specific for activated human protein C (APC) and it lacks reactivity for Protein C. The monoclonal antibody recognizes the light chain of the APC molecule.

Protein C is a protein that plays a crucial role in the regulation of blood clotting. It is a vitamin K-dependent serine protease that is activated by thrombin, a protein that promotes blood clotting. Once activated, protein C proteolytically cleaves and inactivates factors V and VIII, which are essential for the formation of blood clots. This process helps to prevent excessive blood clotting and maintain the normal flow of blood through the body. In addition to its anticoagulant function, protein C has also been shown to have anti-inflammatory and cytoprotective properties, suggesting that it may have a broader role in maintaining vascular health.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Protein C (Active)
Applications Inhibition Assays, Western Blot
Classification Monoclonal
Clone PC107
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA and 0.02% sodium azide
Gene PROC
Gene Accession No. P04070
Gene Alias Active Protein C
Gene Symbols PROC
Host Species Rat
Immunogen Monoclonal antibody PC107 is specific for activated Protein C (APC.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5624
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.