Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

protein C light chain Rabbit anti-Human∣Rat, Polyclonal, Bioss
SDP

Catalog No. 501980904
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0904 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0904 Supplier Bioss Supplier No. BS10486R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Protein C is a vitamin K-dependent plasma glycoprotein that is cleaved to its activated form by the thrombin-thrombomodulin complex. Protein C in its activated form contains a serine protease domain and functions in degradation of the activated forms of coagulation factors V and VIII. Protein C is a vitamin K-dependent serine protease produced in the liver and made up of 2 polypeptide chains. The 62 kDa proenzyme is activated by thrombin bound to an endothelial surface receptor and the active enzyme cleaves factor Va and VIIIa and thus inhibits blood coagulation. The molecular weight of the active enzyme is 55kDa and the normal concentrations in human plasma is approximately 1-3 ng/ml because of the very fast turnover, the proenzyme concentration is approximately 3 μg/ml. Mutations in the Protein C gene have been associated with thrombophilia due to protein C deficiency, neonatal purpura fulminans, and recurrent venous thrombosis.

Specifications

Antigen protein C light chain
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PROC
Gene Accession No. P04070, P31394
Gene Alias Activation peptide; Anticoagulant protein C; APC; Autoprothrombin IIA; Blood coagulation factor XIV; BOS_1536; EDQVDPRLIDGK; inactivator of coagulation factors Va, VIII; PC; prepro-protein C; PROC; PROC1; protein C; protein C (inactivator of coagulation factors Va and VIIIa); protein C, inactivator of coagulation factors Va and VIIIa; THPH3; THPH4; type I protein C; Vitamin K-dependent protein C; Vitamin K-dependent protein C heavy chain; Vitamin K-dependent protein C light chain
Gene Symbols PROC
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Vitamin K-dependent protein C light chain.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25268, 5624
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.