Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Protein Disulfide Isomerase/P4HB Antibody (16), Allophycocyanin, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289676APC
Description
Protein Disulfide Isomerase/P4HB Monoclonal antibody specifically detects Protein Disulfide Isomerase/P4HB in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, ImmunoprecipitationSpecifications
Protein Disulfide Isomerase/P4HB | |
Monoclonal | |
APC | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunocytochemistry, Immunoprecipitation | |
16 | |
Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation | |
Cellular thyroid hormone-binding protein, collagen prolyl 4-hydroxylase beta, DSI, ERBA2L, GIT, glutathione-insulin transhydrogenase, P4Hbeta, p55, PDIA1procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), betapolypeptide (protein disulfide isomerase-associated 1), PDIEC 5.3.4.1, PHDB, PO4DB, PO4HB, procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), betapolypeptide, PROHBprocollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), betapolypeptide (protein disulfide isomerase; thyroid hormone binding protein p55), Prolyl 4-hydroxylase subunit beta, prolyl 4-hydroxylase, beta polypeptide, protein disulfide isomerase family A, member 1, protein disulfide isomerase/oxidoreductase, protein disulfide isomerase-associated 1, protein disulfide-isomerase, protocollagen hydroxylase, thyroid hormone-binding protein p55 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Protein Disulfide Isomerase/P4HB (Accession#: NP_000909.2; Met 1-Lys 505). | |
0.1 mL | |
Biologically Active Proteins, Cancer, Cellular Markers, ER Markers, Fibroblast Cell Markers, Hypoxia | |
5034 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction