Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Protein Disulfide Isomerase/P4HB Mouse Recombinant Protein
SDP

Catalog No. NBP25229725 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
0.25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP25229725 0.25 mg
NBP25229705 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP25229725 Supplier Novus Biologicals™ Supplier No. NBP2522970.25MG

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity

A bioactive recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 20-509 of Mouse Protein Disulfide Isomerase/P4HB The Recombinant Mouse Protein Disulfide Isomerase/P4HB Protein is derived from E. coli. The Recombinant Mouse Protein Disulfide Isomerase/P4HB Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.

Specifications

Concentration 0.5mg/mL
For Use With (Application) SDS-PAGE
Formulation Phosphate buffered saline (pH 7.4) containing 10% glycerol
Gene ID (Entrez) 5034
Molecular Weight (g/mol) TMW: 56.1kDa
Quantity 0.25 mg
Source Baculovirus
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Alias Cellular thyroid hormone-binding protein, collagen prolyl 4-hydroxylase beta, DSI, ERBA2L, GIT, glutathione-insulin transhydrogenase, P4Hbeta, p55, PDIA1procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), betapolypeptide (protein disulfide isomerase-associated 1), PDIEC 5.3.4.1, PHDB, PO4DB, PO4HB, procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), betapolypeptide, PROHBprocollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), betapolypeptide (protein disulfide isomerase; thyroid hormone binding protein p55), Prolyl 4-hydroxylase subunit beta, prolyl 4-hydroxylase, beta polypeptide, protein disulfide isomerase family A, member 1, protein disulfide isomerase/oxidoreductase, protein disulfide isomerase-associated 1, protein disulfide-isomerase, protocollagen hydroxylase, thyroid hormone-binding protein p55
Common Name Protein Disulfide Isomerase/P4HB
Biological Activity Specific activity is >120 A650/cm/min/mg, obtained by measuring the increase of insulprecipitation absorbance at 650nm resulting from the reduction of insulin.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain
Show More Show Less

For research use only and is not approved for use in humans or in clinical diagnosis

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.