Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Protein Disulfide Isomerase/P4HB Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30367820UL
Description
Protein Disulfide Isomerase/P4HB Polyclonal specifically detects Protein Disulfide Isomerase/P4HB in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockout Validated.Specifications
Protein Disulfide Isomerase/P4HB | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, KnockDown | |
Unconjugated | |
Cellular thyroid hormone-binding protein, collagen prolyl 4-hydroxylase beta, DSI, ERBA2L, GIT, glutathione-insulin transhydrogenase, P4Hbeta, p55, PDIA1procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), betapolypeptide (protein disulfide isomerase-associated 1), PDIEC 5.3.4.1, PHDB, PO4DB, PO4HB, procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), betapolypeptide, PROHBprocollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), betapolypeptide (protein disulfide isomerase; thyroid hormone binding protein p55), Prolyl 4-hydroxylase subunit beta, prolyl 4-hydroxylase, beta polypeptide, protein disulfide isomerase family A, member 1, protein disulfide isomerase/oxidoreductase, protein disulfide isomerase-associated 1, protein disulfide-isomerase, protocollagen hydroxylase, thyroid hormone-binding protein p55 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 329-508 of human Protein Disulfide Isomerase/P4HB (NP_000909.2). | |
20 μL | |
Cellular Markers, ER Markers, Fibroblast Cell Markers, Hypoxia | |
5034 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction