Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Protein G Mag Sepharose™ Xtra

Catalog No. 45002512 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
2 x 1 mL
5 x 1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-002-512 2 x 1 mL
45-002-513 5 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-002-512 Supplier Cytiva Supplier No. 28967066
Add to Cart
Add to Cart

High binding capacity, 27mg human IgG/mL medium, for efficient capture of antibody with high yield and purity

  • Non adherant beads
  • Matrix: paramagnetic, spherical, highly cross-linked agarose particles
  • Ligand: Protein G
  • Color: black
  • Operating temperature: room temperature
  • Can be used without detergents
  • For small-scale purification and screening of monoclonal and polyclonal antibodies from serum and cell supernatants
  • Available in 2 x 1mL and 5 x 1mL packs

TRUSTED_SUSTAINABILITY

Specifications

Bead Type Protein G Mag Sepharose Xtra magnetic bead
Binding Capacity 27mg human IgG/mL gel
Matrix Paramagnetic, spherical, highly cross-linked agarose particles
Quantity 2 x 1 mL
Content And Storage 4°C to 8°C
For Use With (Application) For rapid, small-scale purification and screening of monoclonal and polyclonal antibodies from serum and cell supernatants
Ligand Protein G
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.