Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Protein G Polyclonal Antibody, HRP, Rockland™

Rabbit Polyclonal Antibody
$178.35 - $4544.28
Specifications
| Antigen | Protein G |
|---|---|
| Concentration | 1 mg/mL |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | HRP |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-0959
![]()
|
Rockland Immunochemicals
2004031380100 |
100 μg |
Each for $444.03
|
|
|||||
50-199-0960
![]()
|
Rockland Immunochemicals
200403138S |
25 μL |
Each for $178.35
|
|
|||||
50-199-0958
![]()
|
Rockland Immunochemicals
200403138 |
20 mg |
Each for $4,544.28
|
|
|||||
Description
Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Peroxidase, anti-Rabbit Serum as well as purified and partially purified Protein G [Streptococcus species]. Cross reactivity against Protein G from other tissues and species may occur but have not been specifically determined.Specifications
| Protein G | |
| ELISA, Western Blot | |
| HRP | |
| Rabbit | |
| Bacteria | |
| Protein G [Streptococcus species] | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| 1 mg/mL | |
| Polyclonal | |
| Lyophilized | |
| RUO | |
| 0.02M potassium phosphate with 0.15M NaCl, 10 mg/mL BSA and 0.01% gentamicin sulfate; pH 7.2 | |
| Primary | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title