Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Protein Kinase A regulatory subunit I alpha Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$243.00 - $522.00
Specifications
Antigen | Protein Kinase A regulatory subunit I alpha |
---|---|
Applications | Western Blot, Immunohistochemistry, KnockDown |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
Protein Kinase A regulatory subunit I alpha Polyclonal specifically detects Protein Kinase A regulatory subunit I alpha in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockout Validated.Specifications
Protein Kinase A regulatory subunit I alpha | |
Polyclonal | |
Purified | |
RUO | |
Human | |
cAMP-dependent protein kinase regulatory subunit RIalpha, cAMP-dependent protein kinase type I-alpha regulatory chain, cAMP-dependent protein kinase type I-alpha regulatory subunit, CAR, CNC, CNC1, MGC17251, PKA R1 alpha, PKA1, PKR1, PPNAD1, PRKAR1, protein kinase A type 1a regulatory subunit, protein kinase, cAMP-dependent, regulatory, type I, alpha (tissue specificextinguisher 1), Tissue-specific extinguisher 1, TSE1DKFZp779L0468 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human Protein Kinase A regulatory subunit I alpha (NP_002725.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, KnockDown | |
Unconjugated | |
Rabbit | |
Cancer | |
PBS with 50% glycerol, pH7.3. | |
5573 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title