Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Protein Kinase D2 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100636AF350
Description
Protein Kinase D2 Polyclonal antibody specifically detects Protein Kinase D2 in Human samples. It is validated for Western BlotSpecifications
Protein Kinase D2 | |
Polyclonal | |
Western Blot | |
DKFZP586E0820, EC 2.7.11, EC 2.7.11.13, nPKC-D2, PKD2HSPC187, protein kinase D2, serine/threonine-protein kinase D2 | |
The immunogen recognized by this antibody maps to a region between residues 320 and 400. | |
0.1 mL | |
Cancer, Phospho Specific, Protein Kinase, Signal Transduction, Tumor Suppressors | |
25865 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction