Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Protein L Polyclonal Antibody, FITC
GREENER_CHOICE

Catalog No. PA172067
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA172067 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA172067 Supplier Invitrogen™ Supplier No. PA172067
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

PA1-72067 detects Protein L in bacteria samples. The PA1-72067 immunogen is from purified recombinant protein L. Store at -20°C for long term storage.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Protein L
Applications Immunohistochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate FITC
Formulation PBS with 50% glycerol and 0.075% sodium azide
Host Species Chicken
Immunogen Purified recombinant protein L.
Purification Method Protein L
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.