Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Protein L Polyclonal Antibody, HRP

Chicken Polyclonal Antibody

Supplier:  Invitrogen™ PA172069

Catalog No. PA172069


Only null left
Add to Cart

Description

Description

PA1-72069 detects Protein L in bacteria samples. PA1-72069 has been successfully used in ELISA and western blot procedures. The PA1-72069 immunogen is from purified recombinant protein L. Store at -20ºC long term.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Protein L
Polyclonal
HRP
Chicken
Protein L
RUO
Bacteria
Antibody
IgY
ELISA, Western Blot
1 mg/mL
PBS with 50% glycerol and 0.1% chloroacetamide
Purified recombinant protein L.
500 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.