Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ Protein LoBind Tubes

Catalog No. 05412695 Shop All Eppendorf North America Products
Change view
Click to view available options
Capacity (Metric):
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
05-412-695 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-412-695 Supplier Eppendorf™ Supplier No. 2231000344

missing translation for 'validMainframeMsg'

Eppendorf Protein LoBind Tubes are specifically designed for maximum sample recovery in proteomics or other fields of protein research where protein concentration tend to be very small and sample recovery is vital for assay results.

Features:

  • Eppendorf LoBind material guarantees maximum sample recovery for improved assay results
  • Free of surface coating (e.g. silicone) to minimize the risk of sample interference
  • Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors
  • Precise lid sealing to minimize evaporation
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 0.5 mL
For Use With (Application) Preparation or storage of protein, peptide or antibody samples; Enzymatic assays; Storage of virus stok solutions and cell suspensions
No. per Pack 100 (2 bags x 50)
Volume (Metric) 1.5mL
Clean Claims PCR clean
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.