Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Protein mab-21-like 1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$499.50
Specifications
Antigen | Protein mab-21-like 1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
Protein mab-21-like 1 Polyclonal specifically detects Protein mab-21-like 1 in Human samples. It is validated for Western Blot.Specifications
Protein mab-21-like 1 | |
Polyclonal | |
Rabbit | |
Q13394 | |
4081 | |
Synthetic peptides corresponding to MAB21L1(mab-21-like 1 (C. elegans)) The peptide sequence was selected from the N terminal of MAB21L1. Peptide sequence IAAQAKLVYHLNKYYNEKCQARKAAIAKTIREVCKVVSDVLKEVEVQEPR. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
CAGR1mab-21 (C. elegans)-like 1, FLJ10197, mab-21-like 1 (C. elegans), mab-21-like protein 1, protein mab-21-like 1 | |
MAB21L1 | |
IgG | |
41 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title