Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corporation Protein-Pak Hi-Res CM Column

Catalog No. NC9993437 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Change view
Click to view available options
Length (Metric):
100 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
NC9993437 100 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NC9993437 Supplier Waters Corporation Supplier No. 186004929
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Protein-Pak Hi Res Ion-Exchange (IEX) columns were developed to assist in the characterization of recombinant proteins, monoclonal antibodies, and other biological compounds.

  • The non-porous, high compound binding capacity of these particles yields outstanding resolution of charged species in less time compared to use of many traditional porous IEX offerings.
  • In addition, quality control testing with defined protein standards helps ensure consistent batch-to-batch performance.

Specifications

Diameter (Metric) 4.6 mm
Length (Metric) 100 mm
Particle Shape Spherical
Particle Size 7 μm
Quantity 1/Ea.
For Use With (Application) Protein
Product Type Column
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.