Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Protein and Peptide Molecular Weight Markers
GSA_VA

Catalog No. 45000071 Shop All Cytiva Products
Change view
Click to view available options
Product Type:
HMW Native Marker Kit
LMW-SDS Marker Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
45-000-071 HMW Native Marker Kit
45-000-072 LMW-SDS Marker Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-000-071 Supplier Cytiva Supplier No. 17044501
Add to Cart
Add to Cart

Run these markers to determine the molecular weight of uncharacterized protein samples with precision. Cytiva Life Sciences™ Protein and Peptide Molecular Weight Markers work by comparing unknowns with carefully selected standards.

  • Get clear, sharp bands for easy measurement of migration distances
  • Bands can be visualized by using Coomassie™ Brilliant Blue or silver staining

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Description HMW Native Marker Kit
Color White
Form Solid
For Use With (Application) For MW determinations by polyacrylamide gel electrophoresis (PAGE) in homogeneous or gradient gels.
Kit Contents Thyroglobulin: 76μg, 669000 (Mr), porcine thyroid (source), Ferritin: 50μg, 440000 (Mr), equine spleen (source), Catalase: 36μg, 232000 (Mr), bovine liver (source), Lactate dehydrogenase
Molecular Weight (g/mol) Mr 669 000
Quantity 10 Vials
Storage Requirements Unopened vials should be stored at 4°C
Product Type HMW Native Marker Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.